missing translation for 'onlineSavingsMsg'
Learn More

Nitronium Tetrafluoroborate (Technical Grade), TRC

Product Code. 30470839
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470839 50 mg 50mg
30480554 100 mg -
30473524 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470839 Supplier Toronto Research Chemicals Supplier No. TRCN49686850MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
Synonym Nitryl Fluoborate, Nitryl tetrafluoroborate, Nitronium Fluoroborate, Nitronium Tetrafluoroborate, Nitronium Tetrafluoroborate, Nitryl Tetrafluoroborate, Nitryl Tetrafluoroborate
IUPAC Name nitronium;tetrafluoroborate
SMILES F[B-](F)(F)F.O=[N+]=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/BF4.NO2/c2-1(3,4)5;2-1-3/q-1;+1
Formula Weight 132.9958
Chemical Name or Material Nitronium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.