missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 90%

Product Code. p-7050276
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11347818

Brand: Thermo Scientific Alfa Aesar 036374.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

Ingold's method of competitive nitration has been used in order to determine relative reactivities of alkylbenzenes and benzene in homogeneous mononitrations with nitronium tetrafluoroborate in tetramethylene sulfone solutions. Oxidation of both exo-and endo-2-bromonorbornane with nitronium tetrafluoroborate in acetonitrile afforded N-exo-2-norbornylacetamide. Used in the synthesis and characterization of new polyhydroxyorganocarboxylated fullerene derivatives through the electrophilic addition of nitronium tetrafluoroborate onto fullerenes in the presence of arenecarboxylic acid in a nonaqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
PubChem CID 11073463
IUPAC Name nitronium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point 240°C (dec.)
Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula NO2BF4
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 132.81
Percent Purity 90%
Sensitivity Moisture sensitive
Chemical Name or Material Nitronium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.