missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitronium Tetrafluoroborate (0.5 M solution in Sulfolane), TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Nitronium Tetrafluoroborate |
| CAS | 13826-86-3 |
| Molecular Formula | BF4NO2 |
| Quantity | 2.5 g |
| Synonym | Nitryl Fluoborate, Nitryl tetrafluoroborate, Nitronium Fluoroborate, Nitronium Tetrafluoroborate, Nitronium Tetrafluoroborate, Nitryl Tetrafluoroborate, Nitryl Tetrafluoroborate |
| InChI Formula | InChI=1S/BF4.NO2/c2-1(3,4)5;2-1-3/q-1;+1 |
| SMILES | F[B-](F)(F)F.O=[N+]=O |
| IUPAC Name | nitronium;tetrafluoroborate |
| Molecular Weight (g/mol) | 132.81 |
| Formula Weight | 132.9958 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction