missing translation for 'onlineSavingsMsg'
Learn More

Nitron, 95%, Thermo Scientific Chemicals

Product Code. 11483398
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483398 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483398 Supplier Thermo Scientific Alfa Aesar Supplier No. A17809.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2218-94-2
Molecular Formula C20H16N4
Molecular Weight (g/mol) 312.376
MDL Number MFCD00005174
InChI Key CWGBFIRHYJNILV-UHFFFAOYSA-N
Synonym nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
PubChem CID 720071
IUPAC Name (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide
SMILES C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point ∼185°C (decomposition)
Quantity 5 g
Merck Index 14,6613
Formula Weight 312.38
Percent Purity 95%
Chemical Name or Material Nitron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.