missing translation for 'onlineSavingsMsg'
Learn More

Nitromide, MedChemExpress

Product Code. 16418536
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418536 Supplier Medchem Express Supplier No. HYB0945
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nitromide is an anti-parasitic agent.

Nitromide is an anti-parasitic agent.

Specifications

Chemical Name or Material Nitromide
Physical Form Solid
CAS 121-81-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H5N3O5
For Use With (Application) COVID-19-immunoregulation
Synonym 3,5-Dinitrobenzamide
Solubility Information DMSO : ≥ 2.2 mg/mL (10.42 mM)
SMILES O=C(N)C1=CC([N+]([O-])=O)=CC([N+]([O-])=O)=C1
Molecular Weight (g/mol) 211.13
Formula Weight 211.13
Percent Purity 95.79%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.