missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane-d3, for NMR, 99 atom % D, ACROS Organics™

Brand:  Acros Organics 166370050

Code : 29

Additional Details : CAS Number : 13031-32-8 Weight : 0.00590kg Transport : UN number : 1261 Chem class : 3 Pack group : II


Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10104230

  • £47.90 / 5mL


EU Stock 4
Add to basket


Description

Description

For NMR
Specifications

Chemical Identifiers

Nitromethane-d3
13031-32-8
CD3NO2
64.06
99 atom % D
For NMR
CD3NO2
MFCD00044214

Specifications

Nitromethane-d3
98.5% min.
Glass bottle
1.18
100.0°C
5mL
13031-32-8
Liquid
CD3NO2
MFCD00044214
1.1800g/mL
35°C
1.3795
0.1% max. (K.F.)
Clear colorless
99 atom % D
For NMR
CD3NO2
64.06
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nitromethane-d3

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS