missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane-d{3}, 99%(Isotopic), Thermo Scientific Chemicals

Catalog Number 11344428
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13031-32-8
CH3NO2
64.06
MFCD00044214
LYGJENNIWJXYER-FIBGUPNXSA-N
nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms
123293
trideuterio(nitro)methane
[2H]C([2H])([2H])[N+]([O-])=O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13031-32-8
CH3NO2
64.06
MFCD00044214
LYGJENNIWJXYER-FIBGUPNXSA-N
nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms
123293
trideuterio(nitro)methane
[2H]C([2H])([2H])[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.183 g/mL
Odor Characteristic
Refractive Index 1.3795
Quantity 5 g
Boiling Point 100°C
UN Number UN1261
Formula Weight 64.06
Percent Purity 99%
Chemical Name or Material Nitromethane-d3, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'