missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane-d{3}, 99%(Isotopic), Thermo Scientific Chemicals

Product Code. 11334428
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334428 10 g 10g
11344428 5 g 5g
11354428 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11334428 Supplier Thermo Scientific Alfa Aesar Supplier No. 042338.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-32-8
Molecular Formula CH3NO2
Molecular Weight (g/mol) 64.06
MDL Number MFCD00044214
InChI Key LYGJENNIWJXYER-FIBGUPNXSA-N
Synonym nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms
PubChem CID 123293
IUPAC Name trideuterio(nitro)methane
SMILES [2H]C([2H])([2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Color Colorless
Density 1.183g/mL
Odor Characteristic
Refractive Index 1.3795
Quantity 10 g
Boiling Point 100°C
UN Number UN1261
Formula Weight 64.06
Percent Purity 99%
Chemical Name or Material Nitromethane-d3, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.