Learn More
Lenz Laborglasintrumente Nitrogen Round-Bottom Flasks (to Schlenk) with PTFE Needle-Valve Stopcock
Description
- Nitrogen round-bottom flasks (to Schlenk)
- With PTFE needle-valve stopcock
- With conical ground joint and PTFE needle-valve stopcock
- With inscription label, material: DURAN™
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Duran Glass |
| Standard Taper Joint | NS 29/32 |