missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Nitrogen Round-Bottom Flasks (to Schlenk) with PTFE Needle-Valve Stopcock

Product Code. 11913411
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
Standard Taper Joint:
NS 14/23
NS 29/32
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11913411

Brand: Lenz Laborglasintrumente 03360637

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • Nitrogen round-bottom flasks (to Schlenk)
  • With PTFE needle-valve stopcock
  • With conical ground joint and PTFE needle-valve stopcock
  • With inscription label, material: DURAN™

Specifications

Capacity (Metric) 100 mL
Material Duran Glass
Standard Taper Joint NS 14/23
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.