missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurazone, TRC

Product Code. 30479588
Change view
Click to view available options
Quantity:
25 mg
2.5 g
25 g
Unit Size:
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479588 25 mg -
30466573 25 g 25g
30465476 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479588 Supplier Toronto Research Chemicals Supplier No. TRCN49388025MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-87-0
Molecular Formula C6 H6 N4 O4
Molecular Weight (g/mol) 198.14
Synonym 2-[(5-Nitro-2-furanyl)methylene]hydrazinecarboxamide, 1-(5-Nitrofurfurylidene)semioxamazide, (5-Nitro-2-furfurylidenamino)urea, 1-(5-Nitro-2-furfurylidene)semicarbazide, 2-Furancarboxaldehyde, 5-nitro-, semicarbazone, 5-Nitro-2-furaldehyde semicarbazone, 5-Nitro-2-furfural semicarbazone, 5-Nitro-2-furfuraldehyde semicarbazone, 5-Nitrofurazone, 5-Nitrofurfural semicarbazone, Aldomycin, Alfucin, Amifur, Babrocid, Chemofuran, Coxistat, Fura-Septin, FuraZin, Furacilin, Furacilinum, Furacin, Furacine, Furacinetten, Furacoccid, Furacycline, Furalcyn, Furaldon, Furaplast, Furaseen, Furaskin, Furaziline, Furazol W, Furesol, Mammex, Mastofuran, Monafuracin, NFS, NSC 1602, NSC 2100, NSC 44009, Nefco, Nfz mix, Nifucin, Nifuzon, Nitrofural, Nitrofuraldehyde semicarbazone, Nitrofuran, Nitrofuran (bactericide), Nitrofurazone, Nitrozone, Otofural, Topifuran, Vabrocid, Veterinary nitrofurazone
IUPAC Name [(E)-(5-nitrofuran-2-yl)methylideneamino]urea
SMILES NC(=O)N\N=C\c1oc(cc1)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+
Formula Weight 198.0389
Chemical Name or Material Nitrofurazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.