missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurazone 13C,15N2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30323427
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323427 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323427 Supplier Dr. Ehrenstorfer Supplier No. DREA15571030AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrofurazone 13C,15N2
CAS 1217220-85-3
Solvent or Matrix Acetonitrile
Molecular Formula 13C C5 H6 15N2 N2 O4
Quantity 1 mL
Synonym Nitrofurazone-13C,15N2
InChI Formula InChI=1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+/i6+1,8+1,9+1
SMILES N[13C](=O)[15NH]\[15N]=C\c1oc(cc1)[N+](=O)[O-]
IUPAC Name [(E)-(5-nitrofuran-2-yl)methylideneamino]urea
Molecular Weight (g/mol) 201.116
Formula Weight 201.036 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.