missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, Mikromol™

Product Code. 18032300
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032300 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032300 Supplier Mikromol Supplier No. MM0506.00
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nitrofurantoin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrofurantoin
CAS 67-20-9
Analyte or Component Names Nitrofurantoin
Shipping Condition Room Temperature
Molecular Formula C8 H6 N4 O5
Certifications/Compliance ISO 17034
Synonym Nitrofurantoin, 1-[[(5-Nitro-2-furanyl)methylene]amino]-2,4-imidazolidinedione, 1-(5-Nitro-2-furfurylidenamino)hydantoin, 1-(5-Nitro-2-furfurylideneamino)hydantoin, 1-[(5-Nitrofurfurylidene)amino]hydantoin, 5-Nitrofurantoin, Berkfurin, Chemiofuran, Cyantin, Cystit, FuaMed, Fur-ren, Furachel, Furadantin, Furadantine MC, Furadantoin, Furadoin, Furadoine, Furadonin, Furadonine, Furadontin, Furalan, Furantoin, Furantoina, Furatoin, Furina, Furobactina, Furophen T-Caps, Ituran, Macrobid, Macrodantin, N-(5-Nitro-2-furfurylidene)-1-aminohydantoin, N-(5-Nitro-2-furfurylidine)-1-aminohydantoin, N-(5-Nitrofurfurylidene)-1-aminohydantoin, NSC 2107, NSC 44150, Nifurantin, Nitoin, Novofuran, Orafuran, Parfuran, Trantoin, Urantoin, Urizept, Uro-Tablinen, Urodin, Urofurin, Urolong, Welfurin, Zoofurin
InChI Formula InChI=1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+
SMILES [O-][N+](=O)c1oc(\C=N\N2CC(=O)NC2=O)cc1
IUPAC Name 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione
Molecular Weight (g/mol) 238.16
Quantity 500 mg
Formula Weight 238.0338 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.