missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocellulose Membranes, 0.2 μm

Product Code. 10561674 Shop All Thermo Scientific Products
Click to view available options
Dimensions (LxW):
10.5 x 7.9 cm
12 x 8 cm
8 x 8 cm
Sufficient For:
15 Mini-gel Blots
25 Mini-gel Blots
Unit Size:
0.25kg
Pack of 15
This item is not returnable. View return policy

Product Code. 10561674

Brand: Thermo Scientific™ 77012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.2-μm pore size is well-suited for western blotting, and this smaller pore size is ideal for the transfer of low molecular weight proteins or peptides and nucleic acids (<300 bp).

Features include:
100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background—produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 12 cm
Width (Metric) 8 cm
Description Pierce Nitrocellulose Membrane, 0.2 μm, 12 x 8 cm
Quantity 25 Pre-cut Membranes
Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage Store membranes flat at ambient temperature, away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. Keep membranes out of direct sunlight. Sunlight may cause the nitrocellulose to dry and become brittle. Note: Nitrocellulose membrane filters are highly flammable. Keep away from heat and open flame. Flash point is approximately 2°C. Membranes can be sterilized by steam-autoclaving at 121°C.
Shipping Condition Room Temperature
Dimensions (LxW) 12 x 8 cm
Sufficient For 25 Mini-gel Blots
Show More Show Less
compliance-icons
Product Identifier
  • Nitrocellulose Membranes, 0.2
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use water/water spray/water jet/carbon dioxide/sand/foam/alcohol to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Nitrocellulose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.