missing translation for 'onlineSavingsMsg'
Learn More

Nitrocefin (>90%), TRC

Product Code. 30004648
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004648 10 mg 10mg
30006530 100 mg 100mg
30006893 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004648 Supplier Toronto Research Chemicals Supplier No. TRCN49381510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41906-86-9
Molecular Formula C21 H16 N4 O8 S2
Molecular Weight (g/mol) 516.5
Synonym (6R,7R)-3-[(1E)-2-(2,4-dinitrophenyl)ethenyl]-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, [6R-[3(E),6α,7β]]-3-[2-(2,4-dinitrophenyl)ethenyl]-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid;
IUPAC Name (6R,7R)-7-[[(E)-4-(2,4-dinitrophenyl)-2-thiophen-2-ylbut-3-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES OC(=O)C1=CCS[C@@H]2[C@H](NC(=O)C(\C=C\c3ccc(cc3[N+](=O)[O-])[N+](=O)[O-])c4cccs4)C(=O)N12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H16N4O8S2/c26-18(22-17-19(27)23-14(21(28)29)7-9-35-20(17)23)13(16-2-1-8-34-16)6-4-11-3-5-12(24(30)31)10-15(11)25(32)33/h1-8,10,13,17,20H,9H2,(H,22,26)(H,28,29)/b6-4+/t13?,17-,20-/m1/s1
Formula Weight 516.041
Chemical Name or Material Nitrocefin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.