missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nitrobenzene-d5, 99% (Isotopic)

Product Code. 11314428
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11314428 Supplier Alfa Aesar™ Supplier No. 042337.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Physical Form Liquid
Density 1.25g/mL
Flash Point 88°C (190°F)
Quantity 5 g
CAS 4165-60-0
Melting Point 6°C
Boiling Point 211°C
Molecular Formula C6H5NO2
MDL Number MFCD00044415
UN Number UN1662
Synonym nitrobenzene-d5, benzene-d5, nitro, nitrobenzene d5, benzene-d5-, nitro, 1-nitro 2 h? benzene, nitrobenzene-d5, 99.5 atom % d, nitro 2h5 benzene, 1-nitro 2h5 benzene, 2h5 benzene, nitro, nitrobenzene-d5, analytical standard
InChI Key LQNUZADURLCDLV-RALIUCGRSA-N
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])[N+]([O-])=O
IUPAC Name 1-nitro(²H₅)benzene
Molecular Weight (g/mol) 128.14
PubChem CID 123210
Formula Weight 128.14
Percent Purity 99%
Chemical Name or Material Nitrobenzene-d5, Isotopic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.