missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrobenzene D5 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Nitrobenzene D5 |
| CAS | 4165-60-0 |
| Solvent or Matrix | Methanol |
| Molecular Formula | C6 2H5 N O2 |
| Quantity | 1 mL |
| Synonym | Nitrobenzene-D5, Nitrobenzene D5, Benzene-1,2,3,4,5-d5, 6-nitro-, Benzene-d5, nitro- (6CI,7CI,8CI,9CI), 6-Nitrobenzene-1,2,3,4,5-d5, Nitrobenzene-d5, Pentadeuterionitrobenzene, Perdeuteronitrobenzene |
| InChI Formula | InChI=1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])[N+](=O)[O-] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-nitrobenzene |
| Molecular Weight (g/mol) | 128.14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction