missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Reagent Grade, Honeywell™

Product Code. 15647980 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15647980 2.5 L Glass bottle 2.5 L
15637980 1 L Glass bottle 1 L
15627980 10 L Stainless steel can 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15647980 Supplier Honeywell Research Chemicals Supplier No. N109502.5L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 5°C to 6°C
Color Yellow
Density 1.196g/mL at 25°C
pH 8.0 to 8.5 (at 1.0000g/L 20.0°C)
Boiling Point 210°C to 211°C
Explosion Limit 0.4
Flash Point 88°C (190.4°F)
Ignition Point 899°F
Odor Pungent
Packaging Glass bottle
Quantity 2.5 L
UN Number UN1662
Beilstein 507540
Vapor Density 4.2 (vs air)
Formula Weight 123.11g/mol
Vapor Pressure 0.15mmHg (20°C), 50mmHg (120°C)
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.