missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 5.0°C to 6.0°C |
Color | Yellow |
Density | 1.2050g/mL |
Boiling Point | 210.0°C to 211.0°C |
Flash Point | 88°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Nitrobenzene
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H360F-May damage fertility.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : nitrobenzene
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'