missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, ≥99%, ACS Reagent, Honeywell™ Riedel-de Haën™

Product Code. 15608350
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
2 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 L
25mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15608350 25 mL Glass Bottle 25mL
15642740 100 mL Glass Bottle 100mL
15672740 500 mL Glass Bottle 500mL
15652740 1 L Glass Bottle 1 L
15662740 2 L Glass Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15608350 Supplier Honeywell-Riedel-de Haen Supplier No. 25237925ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 5°C to 6°C
Color Yellow
Density 1.196g/mL at 25°C
Boiling Point 210°C to 211°C
Explosion Limit 0.4
Flash Point 88°C (190.4°F)
Ignition Point 899°F
Packaging Glass Bottle
Quantity 25 mL
UN Number UN1662
Beilstein 507540
Vapor Density 4.2 (vs air)
Formula Weight 123.11g/mol
Vapor Pressure 0.15mmHg (20°C), 50mmHg (120°C)
Percent Purity ≥99.0%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.