missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 100.0%, 99.0% purity, For GC analysis, MP Biomedicals™

Product Code. 11459121 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459121 500 mL 500mL
11469121 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459121 Supplier MP Biomedicals Supplier No. 0215585190
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym benzene, nitro, nitro-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Color Yellow
Density 1.196g/mL at 25°C
Assay Percent Range 99%
Quantity 500 mL
Formula Weight 123.12
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.