missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Glass Bottle |
| Analyte or Component Names | Nitrochromic Reagent. |
| Purity Grade Notes | EU Ph Ref: 1060100 |
| Element | Nitrochromic |
| Solvent or Matrix | Nitric Acid |
| Quantity | 200 mL |
| Grade | Eu. Ph. |
Safety and Handling
Product Identifier
- Nitro-Molibdovanadic Reagent. European Pharmacopoeia Ref: 1060100 (RP 140)
- Danger
- Skin corrosion/irritation Category 1 B
- Carcinogenicity Category 2
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
