Learn More
Ansell BioClean™ Durableu™ High Durability Cleanroom Overshoes
Product Code. 15228118
Change view
Click to view available options
Size: 18 in.; Blue
Product Code.
15228118 Size: 18 in.; Blue
1 options
Product Code. 15228118 Supplier Ansell Supplier No. BDBO18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
BioClean Durableu™ disposable overshoes offer superb protection and durability. Resistant to tears, abrasions and a wide range of liquid chemicals, these cleanroom overshoes are low-linting, slip-resistant and have high tensile strength.
  • High Tensile Strength
  • Slip-Resistant
  • Resists tears and abrasions
  • Non-Woven spunbonded Polypropylene/cast polyethylene laminate
  • Manufactured in a facility holding ISO 9001:2008
  • In accordance with ISO 2859
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Polyethylene, Polypropylene
Color Blue
Certifications/Compliance ISO 9001:2015
Packaging Quantity 50 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; eight outer bags per lined carton (400 pieces)