missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-D™ Sterile Disposable Hood

Product Code. 12992255
Change view
Click to view available options
Sterile hood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12992255 Sterile hood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12992255 Supplier Ansell Supplier No. SBDHD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BioClean-D™ hood is a sterile single use hood designed and constructed using three sections for a perfect fit. Made from antistatic durable low-linting CleanTough™ material, the hood features an elasticated face-opening with reinforced edges for exceptional comfort and protection

  • ISO Class 4 and EU GMP Grade A compatible
  • Sterilized by Gamma irradiation
  • Elasticated face opening with reinforced edges
  • Low linting and anti-static property fabric
  • Universal size
  • Low particulate
  • ASTM D6978 05 TESTED
  • HELMKE DRUM TESTED
Applications: Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, industrial

Compliance

ANSI/ISEA Z87.1

TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material Spunbound Polypropylene
Color White
Closure Type Pull-over
Disposable Single-use
Model S-BDHD
For Use With (Application) Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, industrial
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.