missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-D™ Disposable Hood

Product Code. 12962255
Click to view available options
:
Sterile hood
This item is not returnable. View return policy

Product Code. 12962255

Brand: Ansell BDHD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The BioClean-D™ single use three piece hood has been designed and constructed for a perfect fit. Made from antistatic low-linting durable CleanTough™ material, the hood features an elasticated face-opening with reinforced edges for exceptional comfort and protection

  • ISO Class 4 and EU GMP Grade A compatible
  • Sterilized by Gamma irradiation
  • Elasticated face opening with reinforced edges
  • Low linting and anti-static property fabric
  • Universal size
Applications: Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, industrial
TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material Spunbound Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Model BDHD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.