Learn More
Lovibond™ Nitrite LR Tablets
Lovibond Nitrite LR Tablets, Quantity: 100Pack, Physical Form: Tablets
Brand: Lovibond™ 512310BT
Code : NEW
Additional Details : CAS Number : 7681-38-1 Weight : 0.02760kg
Description
Lovibond Nitrite LR Tablets, Quantity: 100Pack, Physical Form: Tablets
Specifications
Tablets | |
100 Pack |
Safety and Handling
- Nitrite LR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P310-Immediately call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium hydrogensulphate, sulphanilic acid