missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, trisodium salt, monohydrate, 99+%

Product Code. 11985331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11985331 2.5 kg Plastic bottle 2.5kg
10498290 25 g Plastic bottle 25g
10408300 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11985331 Supplier Thermo Scientific Acros Supplier No. 147840025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitrilotriacetic acid, trisodium salt, monohydrate, CAS # 18662-53-8, is considered a non-genotoxic compound showing carcinogenic activity and is used as an inducer of tumorigenesis in laboratory studies using animal models. | CAS: 18662-53-8 | C6H6NNa3O6·H2O | 275.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Nitrilotriacetic acid, trisodium salt, monohydrate is considered to be a nongenotoxic compound with carcinogenic activity
• This compound inhibits gap-junctional communication between Chinese hamster lung fibroblasts

Applications

• Nitrilotriacetic acid trisodium salt monohydrate is commonly used in laboratory experiments due to its capacity to induce the tumorigenic process
• It has been used in in vivo experiments as an inducer of bladder and kidney tumors in animal models
• It is used as a chelating agent, which forms coordination compounds with metal ions (chelates)

Literature References
Malcolm, A.; Mills, L. Inhibition of gap-junctional intercellular communication between Chinese hamster lung fibroblasts by di(2-ethylhexyl) phthalate (DEHP) and trisodium nitrilotriacetate monohydrate (NTA). Cell Biology and Toxicology. 1989, 5, 145–153.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18662-53-8
Molecular Formula C6H6NNa3O6·H2O
Molecular Weight (g/mol) 275.09
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Synonym trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Melting Point >320.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula N(CH2CO2Na)3·H2O
Quantity 2.5 kg
Solubility Information Solubility in water: 484g/L (25°C)
Formula Weight 275.09
Percent Purity 99+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Nitrilotriacetic acid, trisodium salt, monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.