missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Nitrification Inhibitor for BOD, Formula 2533™

Product Code. 12066802
Click to view available options
Quantity:
35 g
No. of Tests:
200
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12066802

Brand: Hach™ 253335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate. Use two shots (0.16g) in a 300mL sample to inhibit nitrogenous oxygen demand in the BOD test.

Specifications

Color White
Composition TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate
Description Nitrification inhibitor
Format Powder
For Use With (Application) BOD test
No. of Tests 200
Quantity 35 g
Sample Size 300 mL
Test Method BOD test
Type Formula 2533™
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.