missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ULTREX II Ultrapure Reagent, J.T.Bakertrade;

Product Code. 10128300 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
6/Cs.
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10128300 500 mL 6/Cs. 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10128300 Supplier Avantor J.T.Baker Supplier No. 6901.0500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

CAS 7697-37-2
Melting Point 0°C
Boiling Point 83°C
Density 1.4g/cm3
Packaging 6/Cs.
Assay Percent Range 67 to 70%
Quantity 500 mL
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
UN Number 2031
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information Fully miscible with water
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.