missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 67-70%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™

Product Code. 11395780 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395780 500 mL 500mL
11395800 2.5 L 2.5 L
11395790 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11395780 Supplier Fisher Chemical Supplier No. A509P500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

HNO3, CAS Number-7697-37-2, acidum nitricum, acide nitrique, aqua fortis, azotic acid, salpetersaeure, hydrogen nitrate, nitryl hydroxide, rfna, nital, nitrous fumes, 1L, CHEBI:48107, Colorless Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, Liquid, 944 | CAS: 7697-37-2 | HNO3 | 63.01 g/mol

HNO3, CAS Number-7697-37-2, 500mL, Yellow, 63.02g/mol, -41 deg.C, 11349, Liquid, 0.94kPa (20 deg.C)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1
Color Yellow
Melting Point -41°C
Vapor Pressure 0.94kPa (20°C)
Quantity 500 mL
Physical Form Liquid
Formula Weight 63.02g/mol
compliance-icons compliance-icons
Product Identifier
  • Nitric acid 67-69%
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.