Learn More
Nitric Acid Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Brand: Fisher Chemical J/5550/PB17
26.30 GBP valid until 2022-08-01
Use promo code "17976" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7697-37-2 Weight : 3.55000kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Packaging | HDPE plastic bottle |
---|---|
Quantity | 2.5L |
Chemical Identifiers
7697-37-2 | |
63.012 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
11349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
[N+](=O)(O)[O-] |
Specifications
7697-37-2 | |
11349 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
Liquid | |
100°C | |
Undesignated |
HNO3 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
[N+](=O)(O)[O-] | |
63.012 | |
CHEBI:48107 | |
HDPE plastic bottle | |
1 | |
2.5L |
Safety and Handling
- Nitric acid, 5-20%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.