missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Solstice

Product Code. 16625793
Change view
Click to view available options
Quantity:
35 kg
190 L
Unit Size:
190 L
35kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16625793 190 L 190 L
16635793 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 16625793 Supplier Fluka Supplier No. 30709190L
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid Solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Melting Point -31°C
Boiling Point 122°C (1,013 hPa)
CAS Min % ≥ 50%
CAS Max % <70%
Density 1.400 - 1.480 g/cm3 (20°C)
Quantity 190 L
Linear Formula HNO3
Grade ACS
Chemical Name or Material Nitric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.