missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, puriss. p.a. plus, ≥65% (T), Honeywell Fluka™

Product Code. 15681560 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Poly Bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15681560 250 mL Poly Bottle 250mL
15671560 1 L Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15681560 Supplier Honeywell-Fluka Supplier No. 17078250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a. plus,≥65% (T)

  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Melting Point -31°C
Boiling Point 122°C
Density 1.37 to 1.41g/mL at 20°C
Packaging Poly Bottle
Residue after Ignition ≤0.0002% (as SO4)
Vapor Pressure 8mmHg (20°C)
Quantity 250 mL
Physical Form Liquid
UN Number UN2031
Formula Weight 63.01g/mol
Percent Purity ≥65% (T)
Grade puriss. p.a. plus
Chemical Name or Material Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.