missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Solstice

Product Code. 15663090
Change view
Click to view available options
Quantity:
1 L
2.5 L
40 kg
4 x 2.5 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
Unit Size:
1 L
2.5 L
40kg
Pack of 4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15663090 40 kg Metal Drum 40kg
15633090 1 L Glass Bottle 1 L
15623090 1 L Plastic Bottle 1 L
15653090 2.5 L Glass Bottle 2.5 L
15643090 2.5 L Plastic Bottle 2.5 L
15683090 4 x 2.5 L Glass Bottle Pack of 4
15673090 4 x 2.5 L Poly Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15663090 Supplier Honeywell-Fluka Supplier No. 3070940KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone,≥65%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 122°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.37-1.41 g/mL at 20 °C(lit.)
Packaging Metal Drum
Residue after Ignition ≤0.0005% (as SO4)
Vapor Pressure 8mmHg (20°C)
Quantity 40 kg
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Percent Purity ≥65%
Grade puriss. p.a.
Chemical Name or Material Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitric acid
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.