missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, Environmental Grade, 67-70%, Thermo Scientific Chemicals

Product Code. 15481497
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481497 2.5 L 2.5 L
15471497 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15481497 Supplier Thermo Scientific Alfa Aesar Supplier No. 038747.K4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Main commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Boiling Point 121°C
Density 1.41 g/cm3 at 20°C
Odor Pungent
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 67-70%
Quantity 2.5 L
Physical Form Liquid,single sub-boiling quartz distillation
UN Number 2031
Solubility Information Miscible in water and ether.
Formula Weight 63.01
Grade Environmental
Concentration or Composition (by Analyte or Components) Nitric Acid: 69.0%; Water: 31.0%
Chemical Name or Material Nitric Acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.