missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid concentrate, For 1L standard solution, 0.1 M HNO3 (0.1N), Solstice

Product Code. 15693650
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Packaging:
Ampoule
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15693650 6 x 1 Ea. Ampoule Pack of 6
15683650 1 Ea. Ampoule Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15693650 Supplier Honeywell-Fluka Supplier No. 320426X1EA
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

For 1L standard solution, 0.1 M HNO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 100°C
CAS Min % ≥70.0000%, ≥10.0000%
CAS Max % <90.0000%, <20.0000%
Density 1.065 g/cm3
Packaging Ampoule
Quantity 6 x 1 Ea.
Linear Formula HNO3
Physical Form Concentrate
UN Number UN2031
Formula Weight 63.01
Chemical Name or Material Nitric acid concentrate, For 1L standard solution, 0.1 M HNO3 (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.