missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (Certified ACS Plus), Fisher Chemical

Product Code. 15676804 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Safe-Cote™ Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15676804 2.5 L Safe-Cote™ Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15676804 Supplier Fisher Chemical Supplier No. A200SI212
  • ONLINE EXCLUSIVE PRICE
    £116.00
    £113.00 / 2.5 L
    Save £3.00
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

pH -1.0
Color Colorless Yellow
CAS 7697-37-2, 7732-18-5
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Heavy Metals (as Pb) 0.2ppm max.
Identification Pass Test
Packaging Safe-Cote™ Glass
Residue after Ignition 2ppm max.
Vapor Pressure 0.94 kPa at 20°C
Assay Percent Range 70%
Normality about 15.8
Quantity 2.5 L
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
UN Number 2031
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01
Percent Purity 68.0 to 70.0% (w/w)
Grade Certified ACS Plus
Chemical Name or Material Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.