missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid Blank, VHG™

Product Code. 30003044
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003044 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003044 Supplier VHG Labs Supplier No. VHGHNO3BLK500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nitric Acid Blank
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitric acid
CAS 7697-37-2
Methods ICP/ICP-MS
Analyte or Component Names Nitric acid
Shipping Condition Room Temperature
Molecular Formula H N O3
Synonym Nitric acid
InChI Formula InChI=1S/HNO3/c2-1(3)4/h(H,2,3,4)
SMILES O[N+](=O)[O-]
IUPAC Name nitric acid
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 63.01
Quantity 500 mL
Formula Weight 62.9956 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.