missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, BAKER INSTRA-ANALYZED Plus, J.T. Baker™
522.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
HDPE
Description
BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.
- Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
- Lot to lot consistency ensures reliable results between tests
- Space-saving HDPE packaging with low metallic extractables
- Enhanced safety with tamper-evident caps
- Environmentally-friendly recyclable HDPE bottles
- Strict protocols in an ISO facility ensure quality in manufacturing
Specifications
Color | Colorless |
CAS | 7697-37-2 |
Melting Point | -41.6°C |
Boiling Point | 83°C |
Density | 1.503g/cm3 at 20°C |
Odor | Pungent |
Packaging | HDPE |
Assay Percent Range | 69-69% |
Quantity | 1 L |
Molecular Formula | HNO3 |
Show More |
Safety and Handling
Signal Word
- Danger
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 3
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'