Learn More
Nitric acid, ACS reagent, 68-70% solution in water, ACROS Organics™
Brand: Acros Organics 424000250
Code : 29
Additional Details : CAS Number : 7697-37-2 Weight : 0.08500kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Packaging | Glass Bottle |
---|---|
Quantity | 25mL |
Chemical Identifiers
7697-37-2 | |
63.012 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
[N+](=O)(O)[O-] |
Specifications
Nitric acid | |
25mL | |
30.0 | |
photochemical reactions | |
HNO3 | |
MFCD00011349 | |
15, 6663 | |
0.5ppm max. | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
63.012 | |
CHEBI:48107 | |
Liquid Free From Suspended Matter or Sediment | |
ACS Reagent | |
0.2ppm max. | |
0.0005% max. | |
1.4 | |
1ppm max. | |
122.0°C |
Colorless | |
7697-37-2,7732-18-5 | |
32.0 | |
0.01ppm max. | |
HNO3 | |
01,733; 03,212; 04,356; 05,474 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
Solubility in water: >100 g/100mL (20°C). | |
[N+](=O)(O)[O-] | |
1.4000g/mL | |
944 | |
63.01 | |
68.0 to 70.0% | |
0.2ppm max. | |
Glass Bottle | |
Solubility in water: >100 g/100mL (20°C). | |
this material may darken during storage due to | |
-42.0°C |
Safety and Handling
- Nitric acid, 68-70% solution in water
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.