missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 70%, ULTREX™ II Reagent, J.T. Baker™

Product Code. 15508384 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Fluoropolymer, pre-leached
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15508384 1 L Fluoropolymer, pre-leached Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15508384 Supplier JT Baker Supplier No. 6901.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

For critical elemental analysis with less than 10 parts-per-trillion (ppt) levels. Nitric Acid, 70%, ULTREX™ II Reagent, J.T. Baker™ is an extremely pure acid with low levels of metallic impurities especially for use in ppt trace element analysis.

Purity and consistency are key requirements for all reagent chemicals, but they are especially important with acids.

  • Manufactured using state-of-the-art processing
  • Packaged under clean room conditions
  • Post-packaging characterization is performed using advanced analytical instrumental methods
  • A Certificate of Actual Lot Analysis is provided with every bottle

Specifications

CAS 7697-37-2
Melting Point 0°C
Boiling Point 83°C
Density 1.4g/cm3
Packaging Fluoropolymer, pre-leached
Assay Percent Range 67-70%
Quantity 1 L
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information Fully miscible in water
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01g/mol
Grade Ultrapure reagent
Assay w/w
Chemical Name or Material Nitric acid
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.