missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
20.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
Description
Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.06 g/mL |
Quantity | 500 mL |
Physical Form | Liquid |
Solubility Information | Miscible with water and ether. |
Formula Weight | 63.01 |
Chemical Name or Material | Nitric acid, 2N Standardized Solution |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'