missing translation for 'onlineSavingsMsg'
Learn More

Nitrendipine, Tocris Bioscience™

Product Code. 18752724
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752724 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18752724 Supplier Tocris Bioscience Supplier No. 0601/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CaV1.x blocker

L-type calcium channel blocker. Induces autophagy.
TRUSTED_SUSTAINABILITY

Specifications

CAS 39562-70-4
Molecular Formula C18H20N2O6
Quantity 50 mg
Synonym nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin
InChI Key PVHUJELLJLJGLN-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C
IUPAC Name 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 360.366
PubChem CID 4507
ChEBI CHEBI:7582
Formula Weight 360.37
Percent Purity >99%
Chemical Name or Material Nitrendipine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.