missing translation for 'onlineSavingsMsg'
Learn More

Nitrendipine, MedChemExpress

Product Code. 16442169
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16442169

Brand: Medchem Express HYB0424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a dihydropyridine calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect.

Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a dihydropyridine calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect.

Specifications

Chemical Name or Material Nitrendipine
CAS 39562-70-4
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C18H20N2O6
Quantity 1 g
Synonym BAY-E-5009
Solubility Information DMSO : 50 mg/mL (138.75 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OCC
Molecular Weight (g/mol) 360.36
Formula Weight 360.36
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.