missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitrazine Yellow, indicator grade

Product Code. 10404915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10404915 25 g 25g
10184350 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10404915 Supplier Thermo Scientific Chemicals Supplier No. 190570250
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5424-01-1 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-1-5424
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00003938
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
Synonym 2-(2,4-Dinitrophenylazo)-1-naphthol-3,6-disulfonic acid, disodium salt, 3-(2, 4-Dinitrophenylazo)-4-hydroxy-2, 7-naphthalenedisulfonic acid, 2-(2, 4-Dinitrophenylazo)-1-naphthol-3, 6-disulfonic acid
PubChem CID 72656
IUPAC Name 3-aminopyrazine-2-carboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)N

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV_Vis 85% min.
Visual Transition Interval from pH 6.0(bright yellow) to pH 7.2(bright blue)
Color Brown to Orange
Quantity 25 g
Physical Form Powder
Formula Weight 542.35
Grade Indicator
Chemical Name or Material Nitrazine yellow

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.