missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, Ready To Use, Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Packaging | HDPE plastic bottle |
| CAS | 7757-79-1 |
| Physical Form | Liquid |
| Molecular Formula | KNO3 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| SMILES | [K+].[O-][N+]([O-])=O |
| IUPAC Name | potassium;nitrate |
| Molecular Weight (g/mol) | 101.10 |
| PubChem CID | 24434 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?