missing translation for 'onlineSavingsMsg'
Learn More

Hach™ NitraVer™ 5Nitrate Reagent Powder Pillows

Product Code. 12046666
Click to view available options
Quantity:
100 Pack
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12046666

Brand: Hach™ 2106169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hach™ NitraVer™ 5Nitrate Reagent Powder Pillows are used in the determination of MR and HR nitrate by the cadmium reduction method.

Pre-measured for 10mL samples.

Order Info

dimProductType

Specifications

Analytes NO3-N
Format Powder Pillow
For Use With (Application) Determination of MR and HR nitrate
For Use With (Equipment) All except DR/3000, DR/2010, DR/2000
No. of Tests 100
Quantity 100 Pack
Range 0.3 - 30.0mg/L and 0.1 - 10.0mg/L
Sample Size 10 mL
Storage Requirements 10°C to 25°C
Test Method 8039 HR, 8171 MR - Cadmium reduction
Type Nitrate Reagent Powder Pillow
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitrate Reagent Powder Pillows
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-aminobenzenesulfonic acid, 2,5-dihydroxybenzoic acid, cadmium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.