Learn More
Hach Lange™ Nitrate Cuvette Tests
Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.
Brand: Hach Lange™ LCK339
Additional Details : CAS Number : 7664-93-9 Weight : 0.45000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
- ISO 7890-1-2-1986, DIN 38405 D9-2 certifications
- Shelf life of 23 months from production date
- Storage conditions of 15 - 25 °C
- Measuring ranges of 0.23 - 13.5 mg/L NO3-N and 1.0 - 60.0 mg/L NO3
- Precise and reliable measurement values
- Convenient and error-free dosing of the reagents
Specifications
0.23 to 13.5mg/L NO3-N | |
2.6-Dimethylphenol | |
Nitrate Cuvette Test |
Safety and Handling
- LCK 339 Nitrat/Nitrate sample cuvette
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, phosphoric acid