missing translation for 'onlineSavingsMsg'
Learn More

Nitinol wire, , 0.58mmAlfa Aesar™

Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface

Brand:  Alfa Aesar 044951.H5

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150m

 View more versions of this product

Product Code. 11430940

  • £872.70 / 150m


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Nitinol
Alloy N, straight annealed
Wire, 0.58mm (0.023 in.) dia.
MFCD02091734

Specifications

Nitinol
150m
Silver-gray wire
Wire, 0.58mm (0.023 in.) dia.
Silver-gray
Alloy N, straight annealed
Odorless
MFCD02091734
Videos
SDS