missing translation for 'onlineSavingsMsg'
Learn More

Nitinol tension coil spring, wire diameter 0.75mm (0.030in), mean coil diameter 5.55mm (0.22in), Alloy M

Product Code. p-4824851
Click to view available options
Quantity:
100 Pc.
25 Pc.
5 Pc.
Unit Size:
Pack of 100
Pack of 25
Pack of 5
This item is not returnable. View return policy

Product Code. 15484005

Brand: Thermo Scientific Alfa Aesar 045655.KJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-02-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol tension coil spring
Color Gray
Name Note Wire diameter 0.75mm, mean coil diameter 5.55mm
CAS 7440-02-0, 7440-32-6
Quantity 5 Pc.
Odor Odorless
Form Solid
MDL Number MFCD02091734
Concentration or Composition (by Analyte or Components) Nickel: 55.75%; Titanium: 44.25%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.